Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DTNB SIRNA Assays ›
Gene Symbol:
DTNB
Gene Name:
dystrobrevin beta
Gene Aliases:
-
Chromosome Location:
Chr.2: 25377220 - 25673647 on Build GRCh38
UniGene ID:
Hs.307720
Species:
Human
siRNA ID
HSS102970
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DTNB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256303.1
NP_001243232.1
10
1278
NM_001256304.1
NP_001243233.1
9
1015
NM_001256308.2
NP_001243237.1
9
1149
NM_001320932.1
NP_001307861.1
9
1123
NM_001320933.1
NP_001307862.1
10
1278
NM_001320934.1
NP_001307863.1
10
1278
NM_001320935.1
NP_001307864.1
8
1037
NM_001320936.1
NP_001307865.1
10
1278
NM_021907.4
NP_068707.1
10
1278
NM_033147.3
NP_149159.2
10
1278
NM_033148.3
NP_149160.1
10
1278
NM_183360.2
NP_899204.1
10
1278
NM_183361.2
NP_899205.1
10
1278
XM_005264182.1
XP_005264239.1
10
1195
XM_011532671.1
XP_011530973.1
10
1195
XM_011532672.2
XP_011530974.1
10
1195
XM_011532673.1
XP_011530975.1
10
1195
XM_011532674.1
XP_011530976.1
10
1195
XM_011532675.1
XP_011530977.1
10
1195
XM_011532676.1
XP_011530978.1
10
1195
XM_011532677.2
XP_011530979.1
9
1651
XM_011532679.1
XP_011530981.1
10
1195
XM_011532680.2
XP_011530982.1
9
1116
XM_011532681.2
XP_011530983.1
9
1109
XM_017003529.1
XP_016859018.1
10
1195
XM_017003530.1
XP_016859019.1
10
1195
XM_017003531.1
XP_016859020.1
10
1195
XM_017003532.1
XP_016859021.1
10
1195
XM_017003533.1
XP_016859022.1
10
1195
XM_017003534.1
XP_016859023.1
10
1195
XM_017003535.1
XP_016859024.1
10
1195
XM_017003536.1
XP_016859025.1
10
1195
XM_017003537.1
XP_016859026.1
6
1548
XM_017003538.1
XP_016859027.1
9
1041
XM_017003539.1
XP_016859028.1
9
1052
XM_017003540.1
XP_016859029.1
9
1109
XM_017003541.1
XP_016859030.1
9
1093
XM_017003542.1
XP_016859031.1
9
1047
XM_017003543.1
XP_016859032.1
8
941
XM_017003544.1
XP_016859033.1
9
1047
XM_017003545.1
XP_016859034.1
8
941
GenBank mRNA
AF022728.1
-
10
1176
AF070567.1
-
4
420
AK225724.1
-
9
1026
AK294174.1
-
8
942
AK294357.1
-
9
959
AK300019.1
-
10
1155
AK301386.1
-
9
1151
BC016655.1
-
10
1171
3868207
BC049366.1
-
10
1269
4817552
BT009805.1
-
9
1015
BX389926.1
-
4
398
DQ160290.1
-
9
1015
R15062.1
-
2
128
Y12712.1
-
10
1198
Y15718.1
-
9
1015
Y15722.1
-
9
1015
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Process(es)
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0