Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318503.1 | NP_001305432.1 | 4 | 1195 | ||
NM_001395.3 | NP_001386.1 | 4 | 1141 | |||
XM_011531123.1 | XP_011529425.1 | 4 | 1054 | |||
XM_011531124.1 | XP_011529426.1 | 4 | 1187 | |||
GenBank mRNA | AK314250.1 | - | 4 | 1141 | ||
BC034936.1 | - | 4 | 849 | |||
BC042166.1 | - | 4 | 849 | |||
BC060837.1 | - | 4 | 1195 | 30339218 | ||
U87167.1 | - | 2 | 137 | |||
Y08302.1 | - | 4 | 1050 |