Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LPAR1 SIRNA Assays ›
Gene Symbol:
LPAR1
Gene Name:
lysophosphatidic acid receptor 1
Gene Aliases:
EDG2, GPR26, Gpcr26, LPA1, Mrec1.3, VZG1, edg-2, rec.1.3, vzg-1
Chromosome Location:
Chr.9: 110873252 - 111039259 on Build GRCh38
UniGene ID:
Hs.126667
Species:
Human
siRNA ID
HSS103059
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LPAR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001401.3
NP_001392.2
3
559
NM_057159.2
NP_476500.1
4
637
XM_005251781.3
XP_005251838.1
4
782
XM_005251782.3
XP_005251839.1
4
876
XM_017014383.1
XP_016869872.1
4
676
XM_017014384.1
XP_016869873.1
5
1050
XM_017014385.1
XP_016869874.1
6
643
XM_017014386.1
XP_016869875.1
6
943
XM_017014387.1
XP_016869876.1
6
679
XM_017014388.1
XP_016869877.1
6
862
XM_017014389.1
XP_016869878.1
5
605
XM_017014390.1
XP_016869879.1
6
640
XM_017014391.1
XP_016869880.1
6
664
XM_017014392.1
XP_016869881.1
5
907
XM_017014393.1
XP_016869882.1
6
703
XM_017014394.1
XP_016869883.1
5
832
XM_017014395.1
XP_016869884.1
5
660
XM_017014396.1
XP_016869885.1
4
602
XM_017014397.1
XP_016869886.1
5
533
XM_017014398.1
XP_016869887.1
5
776
XM_017014399.1
XP_016869888.1
5
531
XM_017014400.1
XP_016869889.1
6
892
XM_017014401.1
XP_016869890.1
5
563
XM_017014402.1
XP_016869891.1
5
620
XM_017014403.1
XP_016869892.1
6
618
XM_017014404.1
XP_016869893.1
4
816
XM_017014405.1
XP_016869894.1
6
805
XM_017014406.1
XP_016869895.1
5
539
XM_017014407.1
XP_016869896.1
5
595
GenBank mRNA
AK022808.1
-
3
559
AK290287.1
-
5
912
AK296374.1
-
2
240
AK298590.1
-
2
216
AK299279.1
-
2
290
AY322546.1
-
2
211
BC036034.1
-
4
596
5272431
BC073167.1
-
5
506
6193969
BI461641.1
-
4
605
U78192.1
-
3
333
U80811.1
-
2
237
Y09479.1
-
3
333
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
G-protein coupled receptor signaling pathway
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
activation of phospholipase C activity
positive regulation of cytosolic calcium ion concentration
regulation of cell shape
negative regulation of neuron projection development
oligodendrocyte development
cerebellum development
optic nerve development
corpus callosum development
negative regulation of cAMP biosynthetic process
bleb assembly
positive regulation of Rho protein signal transduction
myelination
positive regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of MAPK cascade
positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
positive regulation of stress fiber assembly
cell chemotaxis
positive regulation of dendritic spine development
cellular response to oxygen levels
positive regulation of smooth muscle cell chemotaxis
cellular response to 1-oleoyl-sn-glycerol 3-phosphate
G-protein alpha-subunit binding
G-protein coupled receptor activity
protein binding
PDZ domain binding
lysophosphatidic acid binding
lysophosphatidic acid receptor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0