Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004093.3 | NP_004084.1 | 2 | 345 | ||
XM_017020406.1 | XP_016875895.1 | 2 | 8606 | |||
GenBank mRNA | BC069342.1 | - | 2 | 272 | 7262269 | |
BC074856.2 | - | 2 | 272 | 30915225 | ||
BC074857.2 | - | 2 | 272 | 30915462 | ||
BC105955.1 | - | 2 | 290 | 40034079 | ||
BC105956.1 | - | 2 | 290 | 40034078 | ||
BC105957.1 | - | 2 | 290 | 40034080 | ||
DR004737.1 | - | 2 | 353 | |||
L38734.1 | - | 2 | 300 | |||
U16797.1 | - | 2 | 276 | |||
U81262.1 | - | 2 | 276 |