Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ELAVL4 SIRNA Assays ›
Gene Symbol:
ELAVL4
Gene Name:
ELAV like neuron-specific RNA binding protein 4
Gene Aliases:
HUD, PNEM
Chromosome Location:
Chr.1: 50048014 - 50203786 on Build GRCh38
UniGene ID:
Hs.213050
Species:
Human
siRNA ID
HSS103177
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ELAVL4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001144774.2
NP_001138246.1
2
441
NM_001144775.2
NP_001138247.2
2
291
NM_001144776.2
NP_001138248.1
2
253
NM_001144777.2
NP_001138249.1
2
331
NM_001294348.1
NP_001281277.1
2
511
NM_001324208.1
NP_001311137.1
2
331
NM_001324209.1
NP_001311138.1
3
528
NM_001324212.1
NP_001311141.1
3
528
NM_001324213.1
NP_001311142.1
2
511
NM_001324214.1
NP_001311143.1
3
598
NM_001324215.1
NP_001311144.1
2
511
NM_001324216.1
NP_001311145.1
2
441
NM_001324217.1
NP_001311146.1
2
511
NM_021952.4
NP_068771.2
2
441
NR_136725.1
3
598
XM_006710411.3
XP_006710474.1
2
423
XM_011540889.2
XP_011539191.1
2
306
XM_011540890.2
XP_011539192.1
2
305
XM_011540893.2
XP_011539195.1
2
253
XM_011540894.2
XP_011539196.1
3
595
XM_011540895.2
XP_011539197.1
3
499
XM_017000538.1
XP_016856027.1
2
425
XM_017000539.1
XP_016856028.1
2
1052
XM_017000540.1
XP_016856029.1
2
309
XM_017000541.1
XP_016856030.1
2
441
XM_017000542.1
XP_016856031.1
2
290
GenBank mRNA
AK297338.1
-
2
331
AK298817.1
-
2
320
AK298902.1
-
2
511
AK309653.1
-
2
286
AY033995.1
-
2
253
AY033996.1
-
2
264
AY033997.1
-
2
428
AY033998.1
-
2
395
BC036071.1
-
2
399
5286347
BM509527.1
-
2
298
DB472040.1
-
2
441
DC351092.1
-
2
331
M62843.1
-
2
265
Back To Top
More Information
Panther Classification:
Molecular Function -
translation initiation factor
Gene Ontology Categories:
Function(s)
Process(es)
RNA processing
mRNA processing
nucleotide binding
RNA binding
mRNA 3'-UTR binding
AU-rich element binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0