Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008707.1 | NP_001008707.1 | 21 | 2300 | ||
NM_004434.2 | NP_004425.2 | 20 | 2243 | |||
XM_005267397.1 | XP_005267454.1 | 21 | 2370 | |||
XM_005267398.3 | XP_005267455.1 | 21 | 2303 | |||
XM_005267399.4 | XP_005267456.2 | 21 | 2342 | |||
XM_005267400.4 | XP_005267457.1 | 20 | 2269 | |||
XM_011536540.2 | XP_011534842.1 | 20 | 2316 | |||
XM_011536541.1 | XP_011534843.1 | 21 | 2383 | |||
XM_011536542.2 | XP_011534844.1 | 20 | 2210 | |||
XM_017021074.1 | XP_016876563.1 | 19 | 2181 | |||
GenBank mRNA | AK126980.1 | - | 22 | 2256 | ||
AK299812.1 | - | 21 | 2579 | |||
BC032541.1 | - | 20 | 2219 | 5533599 | ||
BC033043.1 | - | 20 | 2243 | 4821202 | ||
U97018.1 | - | 20 | 2242 |