Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001424.5 | NP_001415.1 | 4 | 435 | ||
XM_006720864.3 | XP_006720927.1 | 4 | 429 | |||
GenBank mRNA | AK313134.1 | - | 4 | 351 | ||
BC009687.1 | - | 4 | 326 | 3895094 | ||
BF981157.1 | - | 4 | 417 | |||
U52100.1 | - | 3 | 270 | |||
X94770.1 | - | 3 | 217 |