Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193503.1 | NP_001180432.1 | 7, 8 | 1042 | ||
NM_001976.4 | NP_001967.3 | 9, 10 | 1213 | |||
NM_053013.3 | NP_443739.3 | 9, 10 | 1171 | |||
XM_005256521.2 | XP_005256578.1 | 9, 10 | 1599 | |||
XM_011523729.1 | XP_011522031.1 | 9, 10 | 1189 | |||
XM_017024346.1 | XP_016879835.1 | 9, 10 | 1606 | |||
GenBank mRNA | AK300662.1 | - | 9, 10 | 1057 | ||
AK300673.1 | - | 9, 10 | 1007 | |||
AK300709.1 | - | 7, 8 | 1012 | |||
AL359213.1 | - | 2, 3 | 237 | |||
BC017249.2 | - | 9, 10 | 1128 | 4902149 | ||
F26358.1 | - | 1, 2 | 100 | |||
X16504.1 | - | 8, 9 | 1075 | |||
X51957.1 | - | 8, 9 | 1087 |