Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005232.4 | NP_005223.4 | 15, 16 | 2580 | ||
XM_006715880.3 | XP_006715943.1 | 14, 15 | 2208 | |||
GenBank mRNA | AK290351.1 | - | 15, 16 | 2544 | ||
AK313263.1 | - | 15, 16 | 2553 | |||
BC130291.1 | - | 15, 16 | 2564 | 40146322 | ||
BC144156.1 | - | 13, 14 | 2509 | |||
M18391.1 | - | 15, 16 | 2610 | |||
Z27409.1 | - | 11, 12 | 1638 |