Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EYA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301012.1 | NP_001287941.1 | 16 | 1827 | ||
NM_001301013.1 | NP_001287942.1 | 18 | 2007 | |||
NM_004100.4 | NP_004091.3 | 18 | 2131 | |||
NM_172103.3 | NP_742101.2 | 17 | 2062 | |||
NM_172105.3 | NP_742103.1 | 18 | 2131 | |||
XM_005266851.4 | XP_005266908.1 | 18 | 1856 | |||
XM_005266853.4 | XP_005266910.1 | 17 | 1769 | |||
XM_017010368.1 | XP_016865857.1 | 19 | 2376 | |||
XM_017010369.1 | XP_016865858.1 | 18 | 1838 | |||
XM_017010370.1 | XP_016865859.1 | 17 | 1787 | |||
XM_017010371.1 | XP_016865860.1 | 17 | 1772 | |||
XM_017010372.1 | XP_016865861.1 | 16 | 1694 | |||
XM_017010373.1 | XP_016865862.1 | 16 | 1676 | |||
XM_017010374.1 | XP_016865863.1 | 16 | 1993 | |||
XM_017010375.1 | XP_016865864.1 | 16 | 1967 | |||
GenBank mRNA | AJ007993.1 | - | 3 | 239 | ||
AJ007994.1 | - | 3 | 239 | |||
AK295798.1 | - | 16 | 1994 | |||
AK299378.1 | - | 19 | 1921 | |||
AK301950.1 | - | 17 | 2067 | |||
BC014193.2 | - | 10 | 988 | |||
BC041063.1 | - | 17 | 2001 | 5458911 | ||
KU178044.1 | - | 16 | 1604 | |||
KU178045.1 | - | 15 | 1511 | |||
Y17114.1 | - | 18 | 2114 |