Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ESR1 SIRNA Assays ›
Gene Symbol:
ESR1
Gene Name:
estrogen receptor 1
Gene Aliases:
ER, ESR, ESRA, ESTRR, Era, NR3A1
Chromosome Location:
Chr.6: 151654148 - 152103274 on Build GRCh38
UniGene ID:
Hs.208124
Species:
Human
siRNA ID
HSS103376
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ESR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000125.3
NP_000116.2
6
1519
NM_001122740.1
NP_001116212.1
7
1546
NM_001122741.1
NP_001116213.1
7
1503
NM_001122742.1
NP_001116214.1
8
1655
NM_001291230.1
NP_001278159.1
7
1509
NM_001291241.1
NP_001278170.1
7
1500
NM_001328100.1
NP_001315029.1
6
998
XM_006715374.3
XP_006715437.1
8
1655
XM_006715375.3
XP_006715438.1
6
1652
XM_011535543.2
XP_011533845.1
9
3480
XM_011535544.2
XP_011533846.1
7
1664
XM_011535545.2
XP_011533847.1
7
1553
XM_011535547.2
XP_011533849.1
8
1655
XM_011535549.2
XP_011533851.1
4
666
XM_017010376.1
XP_016865865.1
8
4102
XM_017010377.1
XP_016865866.1
9
4161
XM_017010378.1
XP_016865867.1
8
1613
XM_017010379.1
XP_016865868.1
8
1647
XM_017010380.1
XP_016865869.1
7
3971
XM_017010381.1
XP_016865870.1
7
1399
XM_017010382.1
XP_016865871.1
5
719
XM_017010383.1
XP_016865872.1
4
1024
GenBank mRNA
AB307713.1
-
6
1285
AF120105.1
-
2
373
AF258449.1
-
3
851
AF258450.1
-
2
734
AF258451.1
-
3
613
AK293019.1
-
7
1468
AK304600.1
-
3
414
AK304833.1
-
7
1326
AK311064.1
-
1
59
AK313644.1
-
7
1477
BC128573.1
-
6
1390
40128594
BC128574.1
-
6
1390
40128595
BX640939.1
-
6
999
FJ147558.1
-
5
305
FJ169502.1
-
3
275
FJ169503.1
-
4
225
FJ668387.1
-
3
231
FJ712305.1
-
3
205
FJ712307.1
-
5
551
GQ411258.1
-
4
370
JF810888.1
-
6
1285
LC120329.1
-
6
1494
LC120330.1
-
6
1355
M12674.1
-
6
1577
U47678.1
-
1285
X03635.1
-
6
1645
X73067.1
-
3
304
Z75126.1
-
3
228
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
antral ovarian follicle growth
osteoblast development
chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
signal transduction
phospholipase C-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
androgen metabolic process
negative regulation of gene expression
positive regulation of phospholipase C activity
intracellular steroid hormone receptor signaling pathway
intracellular estrogen receptor signaling pathway
response to estradiol
negative regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of sequence-specific DNA binding transcription factor activity
regulation of neuron apoptotic process
response to estrogen
positive regulation of epidermal growth factor receptor signaling pathway
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of glucose import
decidualization
positive regulation of fibroblast proliferation
negative regulation of smooth muscle cell proliferation
positive regulation of epithelial cell proliferation
positive regulation of sequence-specific DNA binding transcription factor activity
Sertoli cell development
Sertoli cell proliferation
uterus development
vagina development
prostate epithelial cord elongation
prostate epithelial cord arborization involved in prostate glandular acinus morphogenesis
regulation of branching involved in prostate gland morphogenesis
mammary gland branching involved in pregnancy
mammary gland alveolus development
epithelial cell proliferation involved in mammary gland duct elongation
positive regulation of ERK1 and ERK2 cascade
protein localization to chromatin
cellular response to estrogen stimulus
cellular response to estradiol stimulus
negative regulation of triglyceride metabolic process
negative regulation of neuron death
negative regulation of production of miRNAs involved in gene silencing by miRNA
baculum development
RNA polymerase II core promoter proximal region sequence-specific DNA binding
core promoter sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription coactivator binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
steroid hormone receptor activity
steroid binding
protein binding
beta-catenin binding
transcription factor binding
zinc ion binding
enzyme binding
nitric-oxide synthase regulator activity
estrogen receptor activity
type 1 metabotropic glutamate receptor binding
protein complex binding
estrogen response element binding
phosphatidylinositol 3-kinase regulatory subunit binding
RNA polymerase II transcription factor activity, estrogen-activated sequence-specific DNA binding
hormone binding
identical protein binding
ATPase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0