Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESRRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282450.1 | NP_001269379.1 | 5 | 826 | ||
NM_001282451.1 | NP_001269380.1 | 5 | 810 | |||
NM_004451.4 | NP_004442.3 | 5 | 813 | |||
XM_017017313.1 | XP_016872802.1 | 4 | 1317 | |||
GenBank mRNA | BC007915.2 | - | 4 | 283 | ||
BC033701.1 | - | 3 | 222 | |||
BC063795.1 | - | 5 | 763 | 6527166 | ||
BC092470.1 | - | 5 | 826 | 30336569 | ||
BC093720.1 | - | 4 | 614 | 7939565 | ||
BC093722.1 | - | 4 | 614 | 7939567 | ||
HQ692867.1 | - | 4 | 592 | |||
L38487.1 | - | 5 | 766 | |||
X51416.1 | - | 946 |