Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000127.2 | NP_000118.2 | 9, 10 | 2647 | ||
GenBank mRNA | AK130054.1 | - | 4, 5 | 1424 | ||
AK313129.1 | - | 9, 10 | 2395 | |||
BC001174.1 | - | 9, 10 | 2040 | 3502232 | ||
S79639.1 | - | 9, 10 | 2525 |