Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F2RL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256566.1 | NP_001243495.1 | 2 | 239 | ||
NM_004101.3 | NP_004092.1 | 2 | 390 | |||
GenBank mRNA | AK298585.1 | - | 2 | 239 | ||
AK312848.1 | - | 2 | 278 | |||
BC093648.1 | - | 2 | 285 | 7939493 | ||
BC093650.1 | - | 2 | 285 | 7939495 | ||
BC095499.1 | - | 2 | 183 | |||
BX537386.1 | - | 2 | 371 | |||
DC419029.1 | - | 2 | 390 | |||
U92971.1 | - | 2 | 330 |