Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000130.4 | NP_000121.2 | 22 | 6254 | ||
XM_017000660.1 | XP_016856149.1 | 21 | 6263 | |||
GenBank mRNA | AK226079.1 | - | 22 | 6172 | ||
M14335.1 | - | 22 | 6185 | |||
M16967.1 | - | 22 | 6199 |