Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FANCF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022725.3 | NP_073562.1 | 1 | 992 | ||
GenBank mRNA | AF181994.1 | - | 1 | 965 | ||
AF181995.1 | - | 1 | 961 | |||
AK001716.2 | - | 1 | 974 | |||
AK023153.1 | - | 1 | 974 | |||
AK223277.1 | - | 1 | 966 | |||
BC035346.1 | - | 1 | 986 | |||
BC047028.1 | - | 1 | 964 | 5538578 | ||
BC063038.1 | - | 1 | 992 | 5239725 | ||
BC093867.1 | - | 1 | 982 | 7939712 | ||
BC101807.1 | - | 1 | 982 | 8069313 | ||
BX640996.1 | - | 1 | 984 |