Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FHL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243878.1 | NP_001230807.1 | 4 | 523 | ||
NM_004468.4 | NP_004459.2 | 5 | 699 | |||
XM_017000675.1 | XP_016856164.1 | 2 | 123 | |||
GenBank mRNA | AF133732.1 | - | 4 | 520 | ||
AK290641.1 | - | 5 | 699 | |||
AK309179.1 | - | 4 | 708 | |||
BC001351.2 | - | 5 | 689 | 2964682 | ||
BC011697.2 | - | 5 | 689 | 3532981 | ||
BC014501.1 | - | 4 | 510 | 4860329 | ||
BT007052.1 | - | 4 | 520 | |||
BX384389.2 | - | 5 | 754 | |||
CR457425.1 | - | 4 | 520 |