Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLNA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110556.1 | NP_001104026.1 | 25 | 4448 | ||
NM_001456.3 | NP_001447.2 | 25 | 4448 | |||
XM_011531127.1 | XP_011529429.1 | 25 | 4443 | |||
XM_011531128.1 | XP_011529430.1 | 25 | 4443 | |||
XM_011531129.1 | XP_011529431.1 | 25 | 4443 | |||
XM_011531130.1 | XP_011529432.1 | 25 | 4443 | |||
XM_011531131.1 | XP_011529433.1 | 24 | 4242 | |||
GenBank mRNA | AB191259.1 | - | 24 | 4171 | ||
AB191260.1 | - | 24 | 4176 | |||
AB371574.1 | - | 24 | 4176 | |||
AB371575.1 | - | 24 | 4171 | |||
AB371576.1 | - | 24 | 4176 | |||
AB371577.1 | - | 24 | 4171 | |||
AB371578.1 | - | 24 | 4171 | |||
AB371579.1 | - | 24 | 4171 | |||
AB593010.1 | - | 24 | 4311 | |||
AK074048.1 | - | 4 | 648 | |||
AK090427.2 | - | 24 | 4237 | |||
AL157419.1 | - | 2 | 212 | |||
GU727643.1 | - | 25 | 4448 | |||
X53416.1 | - | 25 | 4371 |