Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000818.2 | NP_000809.1 | 8 | 1365 | ||
NM_001134366.1 | NP_001127838.1 | 8 | 1365 | |||
GenBank mRNA | AJ251501.1 | - | 5 | 365 | ||
BC126327.1 | - | 8 | 887 | 8992043 | ||
BC126329.1 | - | 8 | 887 | 8992045 | ||
M74826.1 | - | 8 | 1003 | |||
M81882.1 | - | 8 | 914 | |||
X69936.1 | - | 8 | 907 |