Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172713.1 | NP_001166184.1 | 15 | 4354 | ||
NM_002078.4 | NP_002069.2 | 14 | 4288 | |||
XM_005265069.3 | XP_005265126.1 | 16 | 4373 | |||
XM_005265070.3 | XP_005265127.1 | 16 | 4373 | |||
XM_005265071.3 | XP_005265128.1 | 16 | 4373 | |||
XM_005265072.3 | XP_005265129.1 | 15 | 4325 | |||
XM_005265073.1 | XP_005265130.1 | 15 | 4274 | |||
XM_005265074.1 | XP_005265131.1 | 15 | 4274 | |||
XM_005265075.1 | XP_005265132.1 | 13 | 4162 | |||
XM_006713110.2 | XP_006713173.1 | 14 | 4268 | |||
XM_011533598.2 | XP_011531900.1 | 10 | 3747 | |||
XM_017006184.1 | XP_016861673.1 | 16 | 4373 | |||
XM_017006185.1 | XP_016861674.1 | 16 | 4863 | |||
XM_017006186.1 | XP_016861675.1 | 16 | 4042 | |||
XM_017006187.1 | XP_016861676.1 | 14 | 4226 | |||
XM_017006188.1 | XP_016861677.1 | 12 | 4105 | |||
GenBank mRNA | AB209693.1 | - | 15 | 4269 | ||
U31906.1 | - | 12 | 3542 | |||
U41740.1 | - | 14 | 4213 | |||
X82834.1 | - | 14 | 4135 |