Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005302.3 | NP_005293.1 | 2 | 2299 | ||
GenBank mRNA | AF017262.1 | - | 2 | 1267 | ||
AF502281.2 | - | 2 | 1250 | |||
BC040007.1 | - | 2 | 1901 | 5174678 |
Gene Symbol: SEL1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX649006.1 | - | 1 | 7056 |
Set Membership: |
G-protein coupled receptor |