Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002097.2 | NP_002088.2 | 2 | 442 | ||
GenBank mRNA | BC143711.1 | - | 2 | 508 | ||
BP254755.1 | - | 2 | 423 | |||
D32257.1 | - | 2 | 441 | |||
U14134.1 | - | 2 | 242 | |||
U20272.1 | - | 2 | 242 |