Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUCY2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004963.3 | NP_004954.2 | 10 | 1379 | ||
XM_011520631.2 | XP_011518933.1 | 10 | 1277 | |||
GenBank mRNA | BC136544.1 | - | 10 | 1288 | 9020534 | |
BC136545.1 | - | 10 | 1288 | 9020535 | ||
M73489.1 | - | 10 | 1290 | |||
S57551.1 | - | 10 | 1359 |