Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005330.3 | NP_005321.1 | 3 | 581 | ||
GenBank mRNA | BC015537.2 | - | 5 | 673 | 3884290 | |
CR541912.1 | - | 3 | 328 |