Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002115.2 | NP_002106.2 | 14 | 2006 | ||
XM_017009411.1 | XP_016864900.1 | 13 | 2222 | |||
GenBank mRNA | AB208951.1 | - | 12 | 2864 | ||
BC028129.1 | - | 14 | 2006 | 5211860 | ||
U42303.1 | - | 5 | 925 | |||
U51333.1 | - | 14 | 1988 |