Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021958.3 | NP_068777.1 | 2 | 1182 | ||
GenBank mRNA | AB208812.1 | - | 1 | 1901 | ||
AK056326.1 | - | 2 | 1179 | |||
AK127923.1 | - | 2 | 1072 | |||
AK313298.1 | - | 2 | 1069 | |||
BC007294.1 | - | 2 | 1167 | 3349581 | ||
BC033808.1 | - | 2 | 940 | 5184776 | ||
M60721.1 | - | 2 | 1138 |