Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ONECUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004498.2 | NP_004489.1 | 1 | 970 | ||
XM_011521487.2 | XP_011519789.1 | 1 | 1222 | |||
GenBank mRNA | BC140830.1 | - | 1 | 937 | 9021698 | |
EU532019.1 | - | 1 | 590 | |||
U96173.1 | - | 1 | 970 |