Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002145.3 | NP_002136.1 | 2 | 1177 | ||
XM_005257275.4 | XP_005257332.1 | 2 | 746 | |||
GenBank mRNA | BC074805.2 | - | 2 | 1077 | 30915528 | |
BC074806.2 | - | 2 | 1077 | 30915314 | ||
X16665.1 | - | 2 | 1135 |