Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000197.1 | NP_000188.1 | 10, 11 | 868 | ||
XM_011518618.2 | XP_011516920.1 | 13, 14 | 1340 | |||
XM_011518619.2 | XP_011516921.1 | 12, 13 | 1178 | |||
XM_017014671.1 | XP_016870160.1 | 12, 13 | 3252 | |||
XM_017014672.1 | XP_016870161.1 | 12, 13 | 3176 | |||
XM_017014673.1 | XP_016870162.1 | 11, 12 | 3217 | |||
XM_017014674.1 | XP_016870163.1 | 11, 12 | 3147 | |||
XM_017014675.1 | XP_016870164.1 | 11, 12 | 2933 | |||
XM_017014677.1 | XP_016870166.1 | 13, 14 | 3833 | |||
GenBank mRNA | BC034281.1 | - | 10, 11 | 852 | 5168012 | |
CR541894.1 | - | 10, 11 | 820 | |||
KU178130.1 | - | 10, 11 | 820 | |||
KU178131.1 | - | 11, 12 | 998 | |||
KU178133.1 | - | 8, 9 | 749 | |||
KU178134.1 | - | 5, 6 | 497 | |||
U05659.1 | - | 10, 11 | 868 |