Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001314039.1 | NP_001300968.1 | 4 | 610 | ||
NM_001539.3 | NP_001530.1 | 5 | 788 | |||
GenBank mRNA | AJ277242.1 | - | 3 | 396 | ||
AK095541.1 | - | 5 | 1635 | |||
AK289623.1 | - | 5 | 716 | |||
AK298725.1 | - | 4 | 538 | |||
AY186741.1 | - | 5 | 714 | |||
BC008182.1 | - | 5 | 701 | 2900892 | ||
BM469197.1 | - | 5 | 689 | |||
BT007292.1 | - | 4 | 597 | |||
D13388.1 | - | 5 | 714 | |||
L08069.1 | - | 5 | 679 |