Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000621.4 | NP_000612.1 | 2 | 815 | ||
GenBank mRNA | A23337.1 | - | 1 | 291 | ||
AA418604.1 | - | 1 | 306 | |||
AF498982.1 | - | 1 | 84 | |||
AK314132.1 | - | 2 | 572 | |||
BC069356.1 | - | 1 | 163 | 7262350 | ||
BC069576.1 | - | 1 | 163 | 7262362 | ||
BC074848.2 | - | 1 | 111 | 30915554 | ||
BC074849.2 | - | 1 | 111 | 30915332 | ||
BC096839.1 | - | 1 | 85 | 6519217 | ||
M86841.1 | - | 1 | 60 | |||
S71229.1 | - | 1 | 134 | |||
X57830.1 | - | 1 | 229 |
Set Membership: |
G-protein coupled receptor |