Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165946.1 | NP_001159418.1 | 11 | 1259 | ||
NM_001322793.1 | NP_001309722.1 | 21 | 2675 | |||
NM_001322794.1 | NP_001309723.1 | 21 | 2558 | |||
NM_001322795.1 | NP_001309724.1 | 22 | 2857 | |||
NM_001322796.1 | NP_001309725.1 | 21 | 2839 | |||
NM_001322797.1 | NP_001309726.1 | 11 | 1259 | |||
NM_004969.3 | NP_004960.2 | 21 | 2675 | |||
NR_136399.1 | 22 | 2799 | ||||
XM_017016187.1 | XP_016871676.1 | 21 | 2598 | |||
XM_017016188.1 | XP_016871677.1 | 21 | 3192 | |||
XM_017016189.1 | XP_016871678.1 | 22 | 2591 | |||
XM_017016190.1 | XP_016871679.1 | 21 | 2718 | |||
GenBank mRNA | AB209204.1 | - | 12 | 1324 | ||
AK093287.1 | - | 11 | 1231 | |||
AK302578.1 | - | 11 | 1212 | |||
AK312810.1 | - | 21 | 2600 | |||
AK316407.1 | - | 11 | 1231 | |||
BC096336.1 | - | 21 | 2639 | 40008464 | ||
BC096337.1 | - | 21 | 2639 | 40008465 | ||
BC096339.3 | - | 21 | 2639 | 40008468 | ||
BX648462.1 | - | 21 | 2840 | |||
M21188.1 | - | 21 | 2656 |