Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000882.3 | NP_000873.2 | 6 | 774 | ||
GenBank mRNA | AF101062.1 | - | 6 | 553 | ||
AF180562.1 | - | 6 | 612 | |||
BC104982.1 | - | 6 | 706 | 8143985 | ||
BC104984.1 | - | 6 | 706 | 8143987 | ||
M65271.1 | - | 6 | 620 | |||
M65291.1 | - | 6 | 654 |