Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181501.1 | NP_852478.1 | 24 | 3053 | ||
GenBank mRNA | AK300423.1 | - | 24 | 3218 | ||
BC137121.1 | - | 24 | 3052 | 9021118 | ||
BC137122.1 | - | 24 | 3052 | 9021119 | ||
BX648284.1 | - | 2 | 2881 | |||
X68742.1 | - | 23 | 2917 |