Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000419.4 | NP_000410.2 | 22, 23 | 2402 | ||
XM_011524749.1 | XP_011523051.1 | 22, 23 | 2306 | |||
XM_011524750.1 | XP_011523052.1 | 22, 23 | 2306 | |||
GenBank mRNA | AB209552.1 | - | 10, 11 | 1069 | ||
AF098114.1 | - | 19 | ||||
AK315335.1 | - | 22, 23 | 2306 | |||
BC117443.1 | - | 22, 23 | 2300 | 40125994 | ||
BC126442.1 | - | 22, 23 | 2300 | 8992158 | ||
J02764.1 | - | 22, 23 | 2275 | |||
M34480.1 | - | 22, 23 | 2306 | |||
X06831.1 | - | 11, 12 | 1101 |