Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002205.4 | NP_002196.4 | 7 | 818 | ||
GenBank mRNA | AK291570.1 | - | 7 | 763 | ||
AK304696.1 | - | 6 | 641 | |||
AK311051.1 | - | 6 | 570 | |||
AK312414.1 | - | 7 | 818 | |||
AY429549.1 | - | 4 | 207 | |||
BC008786.2 | - | 7 | 803 | 3629647 | ||
M13918.2 | - | 7 | 705 | |||
X06256.1 | - | 7 | 773 |