Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017424.2 | NP_001017424.1 | 7 | 1409 | ||
NM_001017425.2 | NP_001017425.2 | 7 | 1210 | |||
NM_014217.3 | NP_055032.1 | 7 | 1086 | |||
XM_011509522.2 | XP_011507824.1 | 7 | 2612 | |||
XM_011509523.1 | XP_011507825.1 | 7 | 1014 | |||
XM_011509524.2 | XP_011507826.1 | 7 | 1061 | |||
XM_017001248.1 | XP_016856737.1 | 8 | 1797 | |||
XM_017001249.1 | XP_016856738.1 | 8 | 1748 | |||
GenBank mRNA | AF004711.1 | - | 7 | 1194 | ||
AF129399.1 | - | 6 | 996 | |||
AF171068.1 | - | 7 | 1010 | |||
AK123409.1 | - | 6 | 980 | |||
AK291483.1 | - | 7 | 1190 | |||
AK315249.1 | - | 8 | 1176 | |||
AY552980.2 | - | 7 | 1096 | |||
AY552981.1 | - | 6 | 914 | |||
BC069462.1 | - | 7 | 1010 | 7262344 | ||
BC101693.1 | - | 7 | 1055 | 8069199 | ||
BC101695.1 | - | 7 | 1055 | 8069201 | ||
BC143586.1 | - | 7 | 1056 | 9052097 | ||
EF165334.1 | - | 7 | 1143 | |||
EF165335.1 | - | 7 | 1108 | |||
KF182338.1 | - | 7 | 1045 | |||
KF182339.1 | - | 7 | 1019 |