Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004521.2 | NP_004512.1 | 1 | 555 | ||
GenBank mRNA | AK289413.1 | - | 1 | 501 | ||
AK307502.1 | - | 2 | 252 | |||
BC010885.1 | - | 1 | 491 | |||
BC065267.1 | - | 1 | 512 | |||
BC080604.1 | - | 1 | 486 | |||
BC111042.1 | - | 1 | 495 | |||
BC126279.1 | - | 1 | 308 | 8991995 | ||
BC126281.1 | - | 1 | 308 | 8991997 | ||
FJ224324.1 | - | 1 | 398 | |||
X65873.1 | - | 1 | 398 |