Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KNG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000893.3 | NP_000884.1 | 9 | 1338 | ||
NM_001102416.2 | NP_001095886.1 | 9 | 1338 | |||
NM_001166451.1 | NP_001159923.1 | 8 | 1230 | |||
GenBank mRNA | AK223589.1 | - | 9 | 1287 | ||
AK290839.1 | - | 9 | 1237 | |||
AK298440.1 | - | 10 | 1252 | |||
AK303768.1 | - | 9 | 1318 | |||
AK315230.1 | - | 9 | 1290 | |||
BC026253.1 | - | 9 | 1016 | |||
BC060039.1 | - | 9 | 1241 | 4593077 | ||
CB148843.1 | - | 2 | 152 | |||
CD014130.1 | - | 8 | 1192 | |||
K02566.1 | - | 9 | 1153 |