Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021013.3 | NP_066293.2 | 7 | 1268 | ||
XM_011524793.1 | XP_011523095.1 | 8 | 1347 | |||
GenBank mRNA | BC033252.1 | - | 5 | 605 | ||
BC041070.1 | - | 7 | 1287 | 5481838 |