Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005565.3 | NP_005556.1 | 6, 7 | 531 | ||
GenBank mRNA | AK292890.1 | - | 6, 7 | 492 | ||
BC016618.1 | - | 6, 7 | 468 | 4476274 | ||
BT007273.1 | - | 6, 7 | 324 | |||
U20158.1 | - | 6, 7 | 579 |