Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LECT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002302.2 | NP_002293.2 | 4 | 643 | ||
GenBank mRNA | AK314092.1 | - | 4 | 592 | ||
BC093670.1 | - | 4 | 504 | 7939515 | ||
BC101579.1 | - | 4 | 504 | 8069085 | ||
BI825931.1 | - | 3 | 563 | |||
D63521.1 | - | 4 | 643 |