Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002311.4 | NP_002302.2 | 17 | 2556 | ||
NM_013975.3 | NP_039269.2 | 17 | 2556 | |||
XM_005257970.3 | XP_005258027.1 | 17 | 2524 | |||
XM_006721896.3 | XP_006721959.1 | 17 | 2525 | |||
XM_017024624.1 | XP_016880113.1 | 17 | 2720 | |||
GenBank mRNA | AK125853.1 | - | 4 | 2280 | ||
AK300363.1 | - | 17 | 2516 | |||
BC068005.1 | - | 17 | 2531 | 6092747 | ||
BG470103.1 | - | 1 | 39 | |||
U40671.1 | - | 17 | 2517 | |||
X84740.1 | - | 17 | 2499 |