Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ABLIM1 SIRNA Assays ›
Gene Symbol:
ABLIM1
Gene Name:
actin binding LIM protein 1
Gene Aliases:
ABLIM, LIMAB1, LIMATIN, abLIM-1
Chromosome Location:
Chr.10: 114431110 - 114821643 on Build GRCh38
UniGene ID:
Hs.438236
Species:
Human
siRNA ID
HSS106065
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ABLIM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001003407.1
NP_001003407.1
21, 22
2253
NM_001003408.1
NP_001003408.1
22, 23
2337
NM_001322882.1
NP_001309811.1
22, 23
2343
NM_001322883.1
NP_001309812.1
20, 21
2148
NM_001322884.1
NP_001309813.1
20, 21
1953
NM_001322885.1
NP_001309814.1
20, 21
1947
NM_001322886.1
NP_001309815.1
20, 21
1911
NM_001322887.1
NP_001309816.1
19, 20
2007
NM_001322888.1
NP_001309817.1
16, 17
1483
NM_001322889.1
NP_001309818.1
16, 17
1468
NM_001322890.1
NP_001309819.1
15, 16
1405
NM_001322891.1
NP_001309820.1
15, 16
1426
NM_001322892.1
NP_001309821.1
15, 16
1399
NM_001322893.1
NP_001309822.1
15, 16
1420
NM_001322894.1
NP_001309823.1
15, 16
1384
NM_001322895.1
NP_001309824.1
15, 16
1363
NM_001322896.1
NP_001309825.1
14, 15
1321
NM_001322897.1
NP_001309826.1
14, 15
1285
NM_001322898.1
NP_001309827.1
14, 15
1258
NM_001322899.1
NP_001309828.1
14, 15
1279
NM_001322900.1
NP_001309829.1
13, 14
1180
NM_002313.5
NP_002304.3
21, 22
2230
NM_006720.3
NP_006711.3
16, 17
1483
XM_005269824.2
XP_005269881.1
20, 21
2848
XM_005269826.1
XP_005269883.1
18, 19
1719
XM_005269827.4
XP_005269884.1
17, 18
2349
XM_006717837.1
XP_006717900.1
23, 24
2811
XM_006717846.3
XP_006717909.1
18, 19
2454
XM_006717847.1
XP_006717910.1
17, 18
1548
XM_011539801.1
XP_011538103.1
23, 24
2215
XM_011539802.1
XP_011538104.1
23, 24
2128
XM_011539803.1
XP_011538105.1
23, 24
2107
XM_011539805.1
XP_011538107.1
23, 24
2468
XM_017016242.1
XP_016871731.1
25, 26
2311
XM_017016243.1
XP_016871732.1
24, 25
2197
XM_017016244.1
XP_016871733.1
24, 25
2191
XM_017016245.1
XP_016871734.1
23, 24
2301
XM_017016246.1
XP_016871735.1
23, 24
2092
XM_017016247.1
XP_016871736.1
20, 21
1866
XM_017016248.1
XP_016871737.1
20, 21
1866
XM_017016249.1
XP_016871738.1
23, 24
2050
XM_017016250.1
XP_016871739.1
22, 23
2217
XM_017016251.1
XP_016871740.1
22, 23
2008
XM_017016252.1
XP_016871741.1
22, 23
2002
XM_017016253.1
XP_016871742.1
22, 23
1966
XM_017016254.1
XP_016871743.1
22, 23
1945
XM_017016255.1
XP_016871744.1
21, 22
2097
XM_017016256.1
XP_016871745.1
20, 21
2890
XM_017016258.1
XP_016871747.1
16, 17
2229
XM_017016259.1
XP_016871748.1
16, 17
1428
GenBank mRNA
AF005654.1
-
21, 22
2251
AK058007.1
-
11, 12
872
AK093471.1
-
21, 22
1752
AK098277.1
-
16, 17
1488
AK122891.1
-
22, 23
2693
AK297344.1
-
20, 21
2186
AK298709.1
-
14, 15
1162
BC002448.2
-
14, 15
1225
3347450
CR749819.1
-
15, 16
1395
D31883.2
-
21, 22
2055
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
cytoskeleton organization
visual perception
organ morphogenesis
lamellipodium assembly
cilium assembly
actin binding
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-whlt2:80/10.239.162.50:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0