Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002333.3 | NP_002324.2 | 6 | 1799 | ||
XM_005258945.1 | XP_005259002.1 | 6 | 1761 | |||
GenBank mRNA | AB009462.1 | - | 6 | 1932 | ||
AK074751.1 | - | 4 | 1515 | |||
AK126750.1 | - | 2 | 2607 | |||
AK129963.1 | - | 3 | 2203 | |||
AK296536.1 | - | 6 | 1680 | |||
AK316314.1 | - | 5 | 1626 | |||
BC007408.2 | - | 6 | 1728 | 3009711 |