Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000896.2 | NP_000887.2 | 3 | 301 | ||
XM_011528014.2 | XP_011526316.1 | 1 | 53 | |||
GenBank mRNA | AB002454.1 | - | 3 | 301 | ||
AK298675.1 | - | 3 | 281 | |||
D12620.1 | - | 3 | 285 | |||
D12621.1 | - | 3 | 267 |