Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTBR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270987.1 | NP_001257916.1 | 10 | 1223 | ||
NM_002342.2 | NP_002333.1 | 10 | 1418 | |||
XM_005253688.1 | XP_005253745.1 | 9 | 1270 | |||
GenBank mRNA | AK027080.1 | - | 10 | 1283 | ||
AK095418.1 | - | 11 | 1247 | |||
AK128019.1 | - | 2 | 1477 | |||
AK293187.1 | - | 10 | 1162 | |||
BC026262.1 | - | 10 | 1290 | 4703362 | ||
BX648074.1 | - | 2 | 1336 | |||
L04270.1 | - | 10 | 1260 |