Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013257.2 | NP_001013275.1 | 10 | 1304 | ||
NM_005581.4 | NP_005572.2 | 10 | 1304 | |||
GenBank mRNA | BC036745.1 | - | 10 | 1255 | ||
BC050450.1 | - | 10 | 1243 | 5754949 | ||
HE572749.1 | - | 10 | 1238 | |||
X80026.1 | - | 10 | 1244 | |||
X83425.1 | - | 10 | 1260 |