Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003688.1 | NP_001003688.1 | 6 | 1352 | ||
NM_005900.2 | NP_005891.1 | 6 | 1528 | |||
XM_005262992.3 | XP_005263049.1 | 6 | 1321 | |||
XM_006714217.2 | XP_006714280.1 | 6 | 1383 | |||
XM_011531961.1 | XP_011530263.1 | 6 | 1343 | |||
XM_011531962.1 | XP_011530264.1 | 6 | 1388 | |||
XM_011531963.1 | XP_011530265.1 | 6 | 1510 | |||
XM_011531964.1 | XP_011530266.1 | 6 | 1540 | |||
GenBank mRNA | AK096624.1 | - | 2 | 2863 | ||
AK293055.1 | - | 6 | 1654 | |||
BC001878.1 | - | 6 | 1453 | 3546771 | ||
BT007386.1 | - | 5 | 1112 | |||
U54826.1 | - | 6 | 1352 | |||
U57456.1 | - | 5 | 1264 | |||
U59423.1 | - | 6 | 1544 | |||
U59912.1 | - | 6 | 1353 |