Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004988.4 | NP_004979.3 | 3 | 997 | ||
GenBank mRNA | AK314979.1 | - | 3 | 997 | ||
AY148486.1 | - | 3 | 955 | |||
BC017555.1 | - | 3 | 964 | 3922845 | ||
BT009789.1 | - | 1 | 777 | |||
CA428506.1 | - | 1 | 710 |